Learn More
Indene, 90%, tech., stabilized, Thermo Scientific Chemicals
313.92 NOK - 2633.63 NOK
Chemical Identifiers
CAS | 95-13-6, 98-29-3 |
---|---|
Molecular Formula | C9H8 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00003777 |
InChI Key | YBYIRNPNPLQARY-UHFFFAOYSA-N |
Synonym | indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy |
PubChem CID | 7219 |
ChEBI | CHEBI:41921 |
IUPAC Name | 1H-indene |
SMILES | C1C=CC2=CC=CC=C21 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10245210
|
Thermo Scientific Acros
122140050 |
5 g | Glass bottle |
313.92 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10255210
|
Thermo Scientific Acros
122142500 |
250 g | Glass bottle |
972.33 NOK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11994451
|
Thermo Scientific Acros
122140010 |
1 kg | Glass bottle |
2633.63 NOK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-13-6, 98-29-3 | |
116.16 | |
YBYIRNPNPLQARY-UHFFFAOYSA-N | |
7219 | |
1H-indene |
C9H8 | |
MFCD00003777 | |
indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy | |
CHEBI:41921 | |
C1C=CC2=CC=CC=C21 |
Specifications
95-13-6, 98-29-3 | |
White to Yellow | |
181.0°C | |
Authentic | |
C9H8 | |
MFCD00003777 | |
15, 4978 | |
indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy | |
YBYIRNPNPLQARY-UHFFFAOYSA-N | |
1H-indene | |
116.16 | |
CHEBI:41921 | |
90% | |
Crystalline Powder |
-5°C to 3°C | |
0.9900g/mL | |
56°C | |
88% min. (GC) | |
1.5670 to 1.5720 | |
05, 515 | |
0.99 | |
Solubility in water: insoluble. Other solubilities: miscible with most organic solvents,soluble in acetone,miscible with ethanol and ethyl ether | |
C1C=CC2=CC=CC=C21 | |
5 to 10ppm tert-butylcatechol | |
7219 | |
116.16 | |
Technical | |
Indene, tech., 90% |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-393-6
missing translation for 'rtecsNumber' : NK8225000
missing translation for 'tsca' : TSCA