Learn More
Ansell Edmont™ HyFlex™ 11-630 Series Gray Polyurethane Medium Weight Gloves
Yarn structure contains an optimal rate of HPPE. Offers high levels of cut resistance for a safe use in controlling, handling and assembly applications involving contacts with sharp pieces.
Brand: Ansell Edmont™ 11630100
This item is not returnable.
View return policy
Description
- The Lycra™ in the liners of the gloves as well as the polyurethane coating provides excellent flexibility and fit.
- The specific formulation of the polyurethane dipping provides a secure grip on dry or slightly oily pieces.
- The PU formulation also increases abrasion resistance on the gloves’ work surface.
- With strong cut resistance
- Knitted construction
Specifications
Polyurethane | |
Polyurethane | |
10 | |
Dupont™ Kevlar™ aramid fibers | |
Coated-palm | |
High cut resistance |
CE Mark, Standard CATEGORY II EN 388 : 4443 | |
Medium Weight Gloves | |
Knitwrist | |
Nylon/Lycra™/Dyneema™/Cotton/Dupont™ Kevlar™ aramid fiber | |
144 Pairs |
Product Content Correction
Product Title
Ansell Edmont™ HyFlex™ 11-630 Series Gray Polyurethane Medium Weight Gloves > Size: 10