Learn More
Ansell HyFlex™ 11-135 Seamless Knit Polyamide Antistatic Gloves with Coated Fingertips
Gloves are knitted using a seamless, non-stop process with high-tenacity polyamid continuous thread. Antistatic properties protect users from electrostatic discharge.
842.00 NOK
Specifications
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
---|---|
Construction | Seamless Knit |
Coating Material | Polyurethane |
Color | Gray |
Hand | Specific |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16059591
|
Ansell
11135060 |
6 |
842.00 NOK
12 pairs |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
16069591
|
Ansell
11135070 |
7 |
842.00 NOK
12 pairs |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
16079591
|
Ansell
11135080 |
8 |
842.00 NOK
12 pairs |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
16089591
|
Ansell
11135090 |
9 |
842.00 NOK
12 pairs |
Estimated Shipment: 14-07-2025
Log in to see stock. |
|||||
16099591
|
Ansell
11135100 |
10 |
842.00 NOK
12 pairs |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
Description
- Gloves are knitted using a seamless, non-stop process
- Textile liner made of high-tenacity polyamid continuous thread that includes copper
- Seamless, fine stitches enhance user comfort
- Stretchable polyurethane coating on the fingertips for grip capabilities
- Antistatic properties offer the user hand protection against electrostatic discharge
- Low lint for cleanroom applications
Specifications
EN 420:2003 + A1:2009 | |
Polyurethane | |
Specific | |
Anti-static Gloves | |
Unisex | |
11-135 |
Seamless Knit | |
Gray | |
Polyamide | |
113XA | |
Knit | |
13 |