Learn More
Hydrochloric Acid, 1M, Chem Lab™
Product Code. p-8014394
5 L, 5 L
Click to view available options
Quantity:
5 L
10 L
20 L
Unit Size:
10 L
20 L
5 L
For laboratory use, Titrations, ASTM D-4047
Specifications
Appearance Colorless
pH <1
Composition Hydrochloric acid 1mol/L
Concentration 36.461g HCl/l H2O = 1 N (±0.002/20°C)
Density 1.02g/mL
Odor Odorless
Quantity 5 L
Physical Form Liquid
Solubility Information Soluble in Water
compliance-icons
Product Identifier
  • Hydrochloric acid 1 mol/l
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid 1
Product Content Correction
Product Title
Hydrochloric Acid, 1M, Chem Lab™ >