Learn More
HSPB2 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21755-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the superfamily of small heat-shock proteins containing a conservative alpha-crystallin domain at the C-terminal part of the molecule. The protein is expressed preferentially in the heart and skeletal muscle. This protein regulates Myotonic Dystrophy Protein Kinase, which plays an important role in maintece of muscle structure and function.Specifications
HSPB2 | |
Polyclonal | |
Unconjugated | |
HSPB2 | |
heat shock 27kDa protein 2, HSPB2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
161476, 3316 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35878, Q16082 | |
HSPB2 | |
HSPB2 Fusion Protein Ag16146 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |