Learn More
Horst™ HS Model Heating Cable
677.00 NOK - 6410.00 NOK
Product Code | Brand | Model | Length (Metric) Power Cord | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Length (Metric) Power Cord | Price | Quantity & Availability | ||||
10789442
|
Horst™
36004 |
HMSG | 1 m |
6410.00 NOK
Each |
Estimated Shipment: 26-08-2025
Log in to see stock. |
||||
10582502
|
Horst™
20101 |
HS | 0.6m |
677.00 NOK
Each |
Estimated Shipment: 26-08-2025
Log in to see stock. |
||||
10007161
|
Horst™
20105 |
HS | 4m |
1385.00 NOK
Each |
Estimated Shipment: 26-08-2025
Log in to see stock. |
||||