Learn More
Welch Ilmvac™ Hose Connector Adapter
Designed for use with Welch Ilmvac™ Vacuum Pumps.
Brand:  Welch Ilmvac™ 710209
466.78 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10484082
  • 479.00 NOK / Each
Only null left
Estimated Shipment: 22-08-2025
Description
  • Stainless steel hose connector
  • For 12mm tube
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tube
Stainless Steel
DN 25 KF - DN 12
Hose Connector Adapter