Learn More
Nabertherm™ Horizontal High-Temperature Tube Furnace
High-temperature horizontal tube furnaces. High-quality insulation materials made of vacuum-formed fiber plates enable energy-saving operation and a fast heating time due to low heat storage and heat conductivity. Different gas supply systems available.
217045.00 NOK - 404350.00 NOK
Product Code | Brand | Diameter (Metric) Process Tube | Temperature (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Process Tube | Temperature (Metric) | Price | Quantity & Availability | ||||
17308119
|
Nabertherm™
RH281V8AN |
80 mm | 1800°C |
315490.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
17358109
|
Nabertherm™
RH151V6AN |
50 mm | 1600°C |
217045.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
17368109
|
Nabertherm™
RH151V7AN |
50 mm | 1700°C |
243550.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
17378109
|
Nabertherm™
RH151V8AN |
50 mm | 1800°C |
266330.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
17388109
|
Nabertherm™
RH281V6AN |
80 mm | 1600°C |
244030.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
17318119
|
Nabertherm™
RH321V6AN |
120 mm | 1600°C |
308300.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
17328119
|
Nabertherm™
RH321V7AN |
120 mm | 1700°C |
350150.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
17338119
|
Nabertherm™
RH321V8AN |
120 mm | 1800°C |
404350.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
17398109
|
Nabertherm™
RH281V7AN |
80 mm | 1700°C |
282760.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||
Description
- Use: horizontal
- Electrical connection: 3-phase
Specifications
P570 | |
Horizontal |