Learn More
Honeywell™ XC™ Clear FogBan™ Safety Glasses
Click to view available options
:
Blue Frames
Description
- Adjustable temples and lenses with flexible nose piece
- Extra flexible frame of blue plastic
- Easily replaceable lenses - can be changed without replacing the entire frame
- All pressure points are relieved for exceptional all-day comfort
Specifications
Specifications
Certifications/Compliance | EN 166 and EN 170 1F 3-1.2 |
Lens Coating | Anti-fog |
Lens Tint | Clear |
Product Type | Eye and Face Protection Spectacles |
For Use With (Application) | General use and protection against impact (45m/s), Mechanical work (grinding, turning, milling) |
Frame Color | Blue |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |
Frame Material | Plastic |