Learn More
Sperian™ Honeywell™ Clearways Faceshield
Offering both strength and durability with economical replacement visors available in acetate or polycarbonate. Sperian™ Honeywell™ Clearways Faceshields are lightweight and comfortable enough to wear all day.
156.00 NOK - 335.92 NOK
Specifications
No. per Pack 1 Ea.
View More Specs
Products 4
Product Code Brand Product Type Lens Coating Lens Material Price Quantity & Availability  
Product Code Brand Product Type Lens Coating Lens Material Price Quantity & Availability  
11806990
View Documents
Sperian™
1002366
Visor Anti-fog Acetate
335.92 NOK
Each
Estimated Shipment: 30-05-2025
Add to basket
12652733
View accessory products View Documents
Sperian™
1002360
Visor Uncoated Acetate
156.00 NOK
Each
In Stock
Edge
Add to basket
11896980
View Documents
Sperian™
1002353
Visor Uncoated Polycarbonate
261.00 NOK
Each
Estimated Shipment: 22-05-2025
Add to basket
11886980
View Documents
Sperian™
1002346
Browguard with Elastic Headband Uncoated N/A
246.00 NOK
Each
In Stock
Edge
Add to basket
Description
Description
  • Recommended for the agriculture, chemical and forestry industries
  • Protects against hazards including abrasion, airborne particles, biohazards, chemicals, impacts, splashes and more
  • Safe, practical, economical integral visor fitting ensures extra protection
Order Info
Note: The image shows both the Browguard and the Visor. Both pieces must be ordered separately. See Browguard (11886980), and Visors (11806990, 12652733, or 11896980).