Learn More
OHAUS™ Holder Clamp
OHAUS™ Holder Clamp provides secure support for holding glassware at elevated positions.
Brand: OHAUS™ 30392228
1146.44 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Stainless steel is autoclavable and corrosion-resistant
- Aluminum is lightweight yet strong
- Cast iron with black enamel finish is chemical-resistant
- Available in different shapes and sizes to fit your laboratory needs
Specifications
Swivel | |
Holder | |
Clamp, Holder, Swivel, CLC-SWIVLS |
Variable Connection Angle | |
Stainless Steel | |
0 to 19 mm |