Learn More
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Cellophane Sheets
For use with Semi-Dry Transfer Units
Brand: Hoefer™ TE73
This item is not returnable.
View return policy
Description
- Porous Cellophane
- 20 x 35.5 cm
- <50 sheets
Specifications
TE 70 Semi Dry Transfer Unit | |
Porous Cellophane Sheet |
Product Content Correction
Product Title
Hoefer™ TE Blotting Unit Replacement Parts and Accessories: Cellophane Sheets > Cellophane sheets; Porous; 20 x 35.5cm; TE 70