Learn More
Hoefer™ SE 260 or miniVE Vertical Electrophoresis Unit Accessory, Plates
Use with SE 260 or miniVE vertical electrophoresis units. Hoefer™ SE 260 or miniVE Vertical Electrophoresis Unit Accessory, Plates come in a variety of styles for particular needs.
Brand: Hoefer™ SE262GN-5
2133.89 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Size: 10 x 10.5 cm
Specifications
SE 260, MiniVE™ Vertical Electrophoresis Unit | |
Low Fluorescence Notched Plate | |
Glass |