Learn More
Hoefer™ Gel Casters for Mini Vertical Electrophoresis Systems
Easy-to-use casters
Brand: Hoefer™ SE245
3232.22 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
SE245 Dual-Gel Caster
- For use with SE250, SE260, or MiniVE Vertical Electrophoresis Systems
- Suitable for single concentration and gradient gels
- SE235 4-Gel Caster is for use with SE260 or MiniVE Electrophoresis Systems
- SE 275 4-Gel Caster and SE215 10-Gel Caster are for use with SE250 Electrophoresis System
Specifications
SE 250, SE 260 Mini Vertical Gel Electrophoresis Unit | |
Gel Caster | |
Casting cradle, clamp assemblies, sealing gasket set, cams | |
1 to 2 |