Learn More
Cytiva HiPrep™ 16/10 Capto Q
Product Code. 11370292 Shop All Cytiva Products
Click to view available options
:
Bed Volume: 20mL
Specifications
Bed Volume 20 mL
Diameter (Metric) Inner 16 mm
Matrix Capto Q Chromatography Media
compliance-icons
Product Identifier
  • HiPrep 16/10 Capto Q
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol