Learn More
Nickel Electro™ Hinged Gable Lid
View samples better with a semi-transparent lid. Nickel Electro™ Hinged Gable Lid maximizes bath area while using its shape to drip condensation back into the bath.
1175.00 NOK
Specifications
For Use With (Application) | Reducing Water Evaporation, Keeping Samples and Water Free From Contamination and Saves Energy |
---|
Includes: dimForUseWithEquipment
Requires: ForUseWithApplication
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12768888
|
Nickel Electro™
GL1-14 |
14 L Unstirred Baths |
1175.00 NOK
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
11419519
|
Nickel Electro™
GL1-4 |
2 L and 4 L Unstirred Baths |
1175.00 NOK
Each |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||||
Description
- Lightweight
- Semi-transparent
- Drips condensation back into bath
Transparent lid, hinged, high profile, handle
Specifications
Reducing Water Evaporation, Keeping Samples and Water Free From Contamination and Saves Energy |