Learn More
Thermo Scientific™ HERAfreeze™ HFC Series -40°C Ultra-Low Temperature Chest Freezers
Store samples in a versatile and reliable setting with a full range of racking solutions for maximum sample storage.
Description
Store samples in a versatile and reliable setting with Thermo Scientific™ HERAfreeze™ HFC −40°C ultra-low temperature chest freezers. Available in four convenient sizes with a full range of racking solutions these freezers deliver all of the same sample protection features of the HERAfreeze™ HFU-B upright series.
Features:- Easy-to-use microprocessor control panel
- 12-character alphanumeric LED displays setpoints, system status and alarm messages
- Access code to ensure setpoint security
- Heavy gauge, cold-rolled steel exterior construction with a powder coat paint finish that resists chipping and rust
- 5 in. (127mm) foamed-in-place, polyurethane insulation
- Optional built-in chart recorder or data logger for continuous monitoring
- Stainless steel interior
- Easy-to-remove air filter
- Ergonomic, lockable door handle
- Convenient interior covers minimize sample exposure to outside conditions.
- Inner sub-lids protect bulk samples from ambient exposure during single sample retrievals
- Freezer includes RS-232 and 4-20 milliamp analog outputs for independent monitoring devices and recorders
- Heavy duty casters with locking tandem wheel are designed for smooth mobility.
- Warranty varies by country, please contact us for details