Learn More
HBS1L Mouse, anti-Human, CoraLite Plus 488, Clone: 1A10C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68146-100UL
Description
The HBS1L antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HBS1L. This antibody recognizes human antigen. The HBS1L antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
HBS1L | |
1A10C4 | |
CoraLite Plus 488 | |
Mouse | |
Primary | |
Human | |
155 kDa | |
IgG1 |
Immunofluorescence | |
1000 μg/mL | |
DKFZp686L13262, EF 1a, ERFS, HBS1, HBS1 like (S. cerevisiae), HBS1 like protein, HBS1L, HSPC276, KIAA1038 | |
100 μL | |
10767 | |
Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |