Learn More
GRIPAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15806-1-AP-150UL
This item is not returnable.
View return policy
Description
The MW of this protein is 97 kDa, and 15806-1-AP specifically recognizes the 97 kDa protein.
GRASP1 is a neuron-specific guanine nucleotide exchange factor for the Ras family of small G proteins (RasGEF) and is associated with the GRIP/AMPA receptor complex in brain.Specifications
GRIPAP1 | |
Polyclonal | |
Unconjugated | |
GRIPAP1 | |
GRASP 1, GRIP1 associated protein 1, GRIPAP1, KIAA1167 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116493, 54645, 56850 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4V328, Q8VD04, Q9JHZ4 | |
Gripap1 | |
GRIPAP1 Fusion Protein Ag8540 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |