Learn More
Gosselin™ Square HDPE Storage Bottles
Graduated, square-shaped bottles with a carrying handle. Gosselin™ Square HDPE Storage Bottles come with a yellow screw-cap closure.
Brand:  Gosselin™ F2700-12
Product Code. 12364129
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • High-density polyethylene construction
  • Break-resistant
  • Translucent
  • Square-shaped with a carrying handle for easy grasping and carrying
  • Graduated
  • Yellow screw-cap closure
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HDPE
Brown
2.7 L
HDPE
30/Pk.
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Gosselin™ Square HDPE Storage Bottles > Capacity: 2.7L