Learn More
GOLPH3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19112-1-AP-150UL
This item is not returnable.
View return policy
Description
GOLPH3 was initially identified as a peripheral membrane protein localized to the trans-Golgi network, but others reported it to be primarily a mitochondrial protein that regulated the mitochondrial mass through the regulation of the mitochondria-specific phospholipid cardiolipin. GOLPH3 has since been implicated in the target of rapamycin (TOR) signalling pathway. Its overexpression in transfected cells led to and increase in anchorage-independent growth and cell proliferation in vitro. Furthermore, GOLPH3-transfected cells enhanced S6 Kinase activity in response to growth factor stimulation by EGF. Simultaneously, AKT phosphorylation increased in these cells, while these events were abrogated in GOLPH3 siRNA treated cells compared to control cells, indicating the GOLPH3 can enhance signalling through TOR-associated complexes. These results suggest that GOLPH3 is a bona fide oncogene and may be a useful target for therapeutic strategies.Specifications
GOLPH3 | |
Polyclonal | |
Unconjugated | |
GOLPH3 | |
Coat protein GPP34, Golgi phosphoprotein 3, GOLPH3, GOPP1, GPP34, MIDAS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64083, 66629, 78961 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CRA5, Q9ERE4, Q9H4A6 | |
GOLPH3 | |
GOLPH3 Fusion Protein Ag5443 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |