Learn More
Gold Germanium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, Premion™, 99.9997% (metals basis), Thermo Scientific™
6550.00 NOK - 19840.42 NOK
Specifications
Chemical Name or Material | Gold Germanium slug |
---|---|
Name Note | 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length |
Avg. Mol. Wt. or Mol. Wt. Range | Au:Ge; 88:12 wt% |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD02091706 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15463495
|
Thermo Scientific Alfa Aesar
014172.KG |
1 Pc. |
6550.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15473495
|
Thermo Scientific Alfa Aesar
014172.KJ |
5 Pc. | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Gold Germanium slug | |
Au:Ge; 88:12 wt% | |
MFCD02091706 | |
Premion™ | |
≈2.9g/slug |
6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length | |
(metals basis) | |
99.9997% | |
Odorless | |
Slug |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures