Learn More
Glyoxal-bis(2-hydroxyanil), ≥97.0% (from N), Honeywell Fluka™
Indicator for metal titration
5465.44 NOK - 31116.77 NOK
Chemical Identifiers
CAS | 1149-16-2 |
---|---|
Molecular Formula | C14H12N2O2 |
Molecular Weight (g/mol) | 240.26 |
MDL Number | MFCD00002183 |
InChI Key | OMJNMRJMNUREOJ-KOBPNRPCSA-N |
PubChem CID | 5356911 |
SMILES | OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15673940
|
Honeywell-Fluka
33255-25G |
25 g | Glass Bottle |
5465.44 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15683940
|
Honeywell-Fluka
33255-6X25G |
6 x 25 g | Glass Bottle |
31116.77 NOK
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1149-16-2 | |
240.26 | |
OMJNMRJMNUREOJ-KOBPNRPCSA-N | |
OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O |
C14H12N2O2 | |
MFCD00002183 | |
5356911 |
Specifications
203°C to 211°C | |
C14H12N2O2 | |
NONH for all modes of transport | |
OMJNMRJMNUREOJ-KOBPNRPCSA-N | |
6-[(E)-2-(2-hydroxyanilino)ethenyl]iminocyclohexa-2,4-dien-1-one | |
5356911 | |
≥97% (from N) | |
Passes test for suitable for metal definition | |
Glyoxal-bis(2-hydroxyanil) |
1149-16-2 | |
MFCD00002183 | |
2808815 | |
OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O | |
240.26 | |
240.26g/mol | |
Odourless | |
Yellow |
Safety and Handling
missing translation for 'rtecsNumber' : 214-560-0
Product Title
Glyoxal-bis(2-hydroxyanil), ≥97.0% (from N), Honeywell Fluka™