Learn More
GLIS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12678-1-AP-20UL
 View more versions of this product
Product Code. 16870524
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
This gene is a member of the GLI-similar zinc finger protein family and encodes a nuclear protein with five C2H2-type zinc finger domains. This protein functions as both a repressor and activator of transcription and is specifically involved in the development of pancreatic beta cells, the thyroid, eye, liver and kidney. Mutations in this gene have been associated with neonatal diabetes and congenital hypothyroidism (NDH). Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only two have been determined.
Specifications
Specifications
GLIS3
Polyclonal
Unconjugated
GLIS3
GLI similar 3, GLIS family zinc finger 3, GLIS3, Zinc finger protein 515, Zinc finger protein GLIS3, ZNF515
Rabbit
Antigen Affinity Chromatography
RUO
169792, 226075, 680671
-20°C
Liquid
Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6XP49, Q8NEA6
GLIS3
GLIS3 Fusion Protein Ag3366
20 μL
Primary
Human, Rat, Mouse