missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLIS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12678-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a member of the GLI-similar zinc finger protein family and encodes a nuclear protein with five C2H2-type zinc finger domains. This protein functions as both a repressor and activator of transcription and is specifically involved in the development of pancreatic beta cells, the thyroid, eye, liver and kidney. Mutations in this gene have been associated with neonatal diabetes and congenital hypothyroidism (NDH). Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only two have been determined.Specifications
GLIS3 | |
Polyclonal | |
Unconjugated | |
GLIS3 | |
GLI similar 3, GLIS family zinc finger 3, GLIS3, Zinc finger protein 515, Zinc finger protein GLIS3, ZNF515 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
169792, 226075, 680671 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6XP49, Q8NEA6 | |
GLIS3 | |
GLIS3 Fusion Protein Ag3366 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction