Learn More
Rogo Sampaic™ Class B Glass Volumetric Pipettes
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Rogo Sampaic™ Class B Glass Volumetric Pipettes are manufactured from clear soda lime glass.
9.00€ - 43.00€
Specifications
Calibrated Yes
Class B
Graduated Yes
View More Specs
Products 6
Product Code Brand Volume (Metric) Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Color Price Quantity & Availability  
11752674
View Documents
Rogo Sampaic™
DGG012
2 mL Black
9.00€
Each
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
11712684
View Documents
Rogo Sampaic™
DGG006
25 mL Black
14.00€
Each
Estimated Shipment: 16-06-2025
to see stock.
Add to basket
11702684
View Documents
Rogo Sampaic™
DGG005
20 mL Yellow
16.00€
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
11752684
View Documents
Rogo Sampaic™
DJR004
10 mL Red
43.00€
Pack of 2
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
11722684
View Documents
Rogo Sampaic™
DGG007
50 mL Orange
16.00€
Each
In Stock
to see stock.
Edge
Add to basket
11792674
View Documents
Rogo Sampaic™
DGG004
10 mL
11.00€
Each
Estimated Shipment: 13-05-2025
Description
  • With vitrified amber graduations
  • With safety bulb
Specifications
Specifications
Yes
B