Learn More
Macherey-Nagel™ Glass Capillary
Glass capillaries with 1 μL volume (approx. 6.5 cm length).
Brand: Macherey-Nagel™ 814022
1023.64 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Glass capillaries with 1 μL volume (approx. 6.5 cm length).
Glass capillaries with 1 μL volume (approx. 6.5 cm length).
Specifications
Capillary Column | |
1 Each |