Learn More
Ansell™ Gladiator™ 16-650 Series Heavy Weight Gloves
Designed for light-to-medium stress glass handling. Crinkle finish provides excellent wet and dry grip on slippery surfaces but also protects the hand from sharp edges.
1965.00 NOK
Specifications
Coating Material | Natural Rubber Latex |
---|---|
Latex Free | No |
Product Type | Heavyweight Gloves |
Style | 16-650 |
Glove Cuff Color | Green |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12670772
|
Ansell™
16650080 |
8 |
1965.00 NOK
12 pairs |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||||
12650772
|
Ansell™
16650100 |
10 |
1965.00 NOK
12 pairs |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||||
12660772
|
Ansell™
16650070 |
7 |
1965.00 NOK
12 pairs |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||||
12680772
|
Ansell™
16650090 |
9 |
1965.00 NOK
12 pairs |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||||
Description
- Crinkle finish for grip on slipper surfaces and protection from sharp edges
- Natural rubber coating
- Cut-and-sewn construction
- Interlock cotton liner
- Fully-coated finish
- Gauntlet cuff
Specifications
Natural Rubber Latex | |
Heavyweight Gloves | |
Green | |
Interlock Cotton | |
Good cushioning, good cut resistance |
No | |
16-650 | |
Gauntlet | |
12 Pairs | |
16-650 |