Learn More
OHAUS™ Frontier™ 5000 Multi-Pro Series Rack
Offering flexibility to maximize sample preparation.
1285.00 NOK - 3905.00 NOK
Specifications
Type Rack
Material Polypropylene
Disposable Reusable
Autoclavable Not Autoclavable
No. per Pack 2 Pieces
View More Specs
Products 18
Product Code Brand Capacity Product Line Price Quantity & Availability  
Product Code Brand Capacity Product Line Price Quantity & Availability  
30130238
View Documents
OHAUS™
83041038
18 x 1.1 mL to 5 mL Sar
3615.00 NOK
Pack of 2
30130256
View Documents
OHAUS™
30772866
1 x 250 mL FA
3905.00 NOK
Pack of 2
30130240
View Documents
OHAUS™
83041017
1 x 100 mL RB V1
1285.00 NOK
Pack of 2
30130251
View Documents
OHAUS™
83041027
7 x 15 mL RB/Vac 1
1285.00 NOK
Pack of 2
30130242
View Documents
OHAUS™
83041028
8 x 7 mL RB/Vac
1285.00 NOK
Pack of 2
30130246
View Documents
OHAUS™
83041020
1 x 50 mL RB V2
1285.00 NOK
Pack of 2
30130235
View Documents
OHAUS™
83041022
1 x 50 mL Rim V3
1285.00 NOK
Pack of 2
30130252
View Documents
OHAUS™
83041026
3 x 15 mL FA
1285.00 NOK
Pack of 2
30130230
View Documents
OHAUS™
83041016
1 x 200 mL FA
1285.00 NOK
Pack of 2
30130228
View Documents
OHAUS™
83041035
1 x 145 mL FB
2305.00 NOK
Pack of 2
30130261
View Documents
OHAUS™
83041021
1 x 50 mL FA V2
1285.00 NOK
Pack of 2
30130226
View Documents
OHAUS™
83041023
1 x 30 mL Rim V2
1285.00 NOK
Pack of 2
30130241
View Documents
OHAUS™
83041025
4 x 15 mL FA
1285.00 NOK
Pack of 2
30130249
View Documents
OHAUS™
83041018
1 x mL RB V2
1285.00 NOK
Pack of 2
30130225
View Documents
OHAUS™
83041024
1 x 30 mL RB
1285.00 NOK
Pack of 2
30130233
View Documents
OHAUS™
83041029
4 x 5 mL FA
1285.00 NOK
Pack of 2
30130265
View Documents
OHAUS™
83041039
1 x 250 mL FB
3615.00 NOK
Pack of 2
30130267
View Documents
OHAUS™
83041019
1 x 50 mL RB V3
1285.00 NOK
Pack of 2
Description
Description
Frontier™ 5000 Rotors
Frontier™ rotors extend the functionality of Frontier™ centrifuges by increasing the number of tubes of varied types and sizes that can be used in conjunction with each of the centrifuge models. Easy to clean and operate, the rotors are durably constructed to withstand demanding centrifugation.
Offering Flexibility to Maximize Sample Preparation
  • Choose from a range of rotors to be used with Frontier™ centrifuges to match capacities and flexibility required for your basic to high-performance sample preparation needs.
  • Designed with usability in mind, the Frontier™ rotors can be effortlessly installed or removed in a few quick and simple steps, and users have easy access to the samples.
  • The sturdy rotors are constructed of high-quality materials and built to last through demanding workflows from autoclave sterilization to sub-zero temperature.
  • OHAUS Frontier™ centrifuges are reliable, versatile and ideal for numerous laboratory applications.
  • Features include small to medium capacity, various speed ranges and non-refrigerated or refrigerated options to suit all requirements.
BIOSEALED Rotors and Buckets
Bio-containment or aerosol tight accessories such as Bioseal Angle rotors for micro tubes and Bioseal buckets used with Swing out rotors for larger sample volumes are submitted for rigorous testing by the German Institute for Standardization (DIN) and as such meet the pertinent international standard, DIN EN 61010-2-020:03-2007.
Specifications
Specifications
Rack
Reusable
2 Pieces