Learn More
Thermo Scientific Chemicals Florisil™, 100-200 mesh, for column chromatography
1740.00 NOK - 20585.00 NOK
Specifications
Particle Size | 100/120 Mesh |
---|---|
Packaging | Glass Bottle |
Material | Florisil |
For Use With (Application) | Column Chromatography |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10328630
|
Thermo Scientific Chemicals
205442500 |
250 g |
1740.00 NOK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10104980
|
Thermo Scientific Chemicals
205445000 |
500 g |
3065.00 NOK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10318903
|
Thermo Scientific Chemicals
205440050 |
5 kg |
20585.00 NOK
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
100/120 Mesh | |
Glass Bottle | |
Florisil | |
Column Chromatography |