Learn More
Heidolph instruments Flex-coupling
Flexible stirrer coupling for 6mm-10mm shafts that are ideal in research, scientific, laboratories and other environments.
Brand:  Heidolph instruments 509-03000-00
Product Code. 10478971
  • 2175.00 NOK / Each
Estimated Shipment: 17-06-2025
Explore more special offers
Description
  • Flexible stirrer coupling for 6mm-10mm shafts
  • Includes clamping stud for stirrer shaft for use with up to 10mm diameter shafts
  • Overhead stirrer accessories
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6-10 mm