Learn More
Grant Instruments™ Flat Platform With Non-Slip Mat for PSU20i
Accommodates low profile containers such as Petri dishes. Grant Instruments™ Flat Platform with Non-Slip Mat includes black rubber pads that provide a nonslip surface for vessels on the platform.
Brand: Grant Instruments™ PP-20
1895.20 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with PSU 10i Orbital Shaking Platform
- Accommodates low profile vessesl such as Petri dishes
- Black rubber pads provide nonslip surface for vessels on the platform.
Specifications
Rubber | |
Flat platform | |
Flat platform |
480 x 380 mm | |
PSU-20i |