Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: C18
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.
220.00 € - 801.00 €
Specifications
Material | Polypropylene |
---|---|
Type | C18 |
Resolution | 40 to 60 μm |
Column Type | C18 |
For Use With (Application) | Non-Polar Compounds, Ideal for Peptides |
Product Code | Brand | Includes | Bed Weight (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Bed Weight (Metric) | Price | Quantity & Availability | ||||
16380166
|
BUCHI
140000053 |
2 Pc. | 120 g |
595.00 €
Pack of 2 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||
16340166
|
BUCHI
140000049 |
5 Pc. | 12 g |
369.00 €
Pack of 5 |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||
16390166
|
BUCHI
140000054 |
1 Pc. | 220 g |
801.00 €
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||
16360166
|
BUCHI
140000051 |
2 Pc. | 40 g |
220.00 €
Pack of 2 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||
16330166
|
BUCHI
140000048 |
5 Pc. | 4 g |
230.00 €
Pack of 5 |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||
16370166
|
BUCHI
140000052 |
2 Pc. | 80 g |
573.00 €
Pack of 2 |
Estimated Shipment: 18-07-2025
Log in to see stock. |
||||
Description
- Most economical solution without sacrificing quality for standard separations
- Large selection of cartridge sizes from 4g-5,000g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
- Silica is ideal for basic applications and polar compounds using organic solvents
- Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
- C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
- Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
- Neutral Alumina is ideal for aldehydes, ketones, and esters
Specifications
Polypropylene | |
40 to 60 μm | |
Non-Polar Compounds, Ideal for Peptides |
C18 | |
C18 | |
Any Flash System |