Learn More
Invitrogen™ eBioscience™ Fixation/Permeabilization Diluent
Intended to be used with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67, as well as cytokines and chemokines.
Brand: Invitrogen™ 00-5223-56
438.18 NOK valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Alert:
Contains Sodium azide
Description
- This must be used in combination with the Fixation/Permeabilization Concentrate (cat. 00-5123) and the Permeabilization Buffer (10X) (cat. 00-8333)
- Alternatively, all buffers required for staining with these antibodies can be purchased as a set in the Foxp3/Transcription Factor Buffer Staining Set (cat. 00-5523)
Specifications
100 mL | |
Fixation Diluent, Permeabilization Diluent | |
eBioscience™ | |
Fixation/Permeabilization Diluent |