Learn More
Fisherbrand™ Amber Soda Glass Wide Mouth Packer Bottles with White Polypropylene/Polyvinyl Lined Cap
Ideal for storage of dry powders.
185.96 NOK - 5344.85 NOK
Specifications
Color | Amber |
---|---|
Includes | Certificate of analysis |
Material | Soda Lime Glass |
Closure Color | White |
Closure Included | Yes |
Product Code | Brand | Capacity (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Quantity | Price | Quantity & Availability | ||||
11724259
|
Fisherbrand™
02911852A |
6 mL | 720/PK. | ||||||
11734259
|
Fisherbrand™
02911799A |
15 mL | 624/Cs. | ||||||
11754319
|
Fisherbrand™
02911432A |
30 mL | 24/Cs. | ||||||
11758749
|
Fisherbrand™
02911433A |
60 mL | 24/Cs. | ||||||
11744259
|
Fisherbrand™
02911448A |
60 mL | 216/Cs. | ||||||
11764319
|
Fisherbrand™
02911434 |
125 mL | 24/Cs. | ||||||
11754259
|
Fisherbrand™
02911449A |
125 mL | 180/Cs. | ||||||
11774319
|
Fisherbrand™
02911435A |
250 mL | 24/Cs. | ||||||
11784319
|
Fisherbrand™
02911436 |
500 mL | 12/Cs. | ||||||
11734319
|
Fisherbrand™
02911451A |
500 mL | 60/Cs. | ||||||
11744319
|
Fisherbrand™
02911452A |
1000 mL | 36/Cs. | ||||||
11778849
|
Fisherbrand™
02911437A |
1000 mL | 12/Cs. | ||||||
Description
- Wide mouth for bottles allows easy accessibility for removal of contents
- Custom cleaned containers and caps are free of visible particulates done using deionised water
- Certified clean containers are cleaned as per EPA guidelines for semi-volatiles, pesticides, PCBs and metals
Specifications
Amber | |
Soda Lime Glass | |
Yes | |
Wide Mouth Packer Bottle | |
Screw Cap |
Certificate of analysis | |
White | |
Round | |
Polypropylene |