Learn More
Ferric Ammonium Sulphate R2 Solution According to European Pharmacopoeia (EP), Reagecon
Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Description
- European Pharmacopoeia Reagent, Solution
- High-purity stable product developed and tested for European pharmacopoeia (EP) applications.
- Used for the preparation of other reagents (4.1.1), manufacture of containers (3.1), and as a reagent in the performance of tests outlined in several EP monographs
- This Reagecon EP reagent is accurate, stable and reproducible.
- Stability is proven (either opened or unopened) for the shelf life of this product.
Specifications
Specifications
Packaging | HDPE Bottle GL45 |
Physical Form | Liquid |
Technique | EP Testing |
Quantity | 1 L |