Learn More
Thermo Scientific™ FastRuler DNA Ladders, ready-to-use
Thermo Scientific FastRuler DNA Ladders are engineered for fast and convenient sizing and approximate quantification of double stranded DNA fragments.
700.00 NOK - 820.00 NOK
Specifications
Gel Type | Agarose |
---|---|
Ready to Load | Yes |
Gel Compatibility | Agarose Gels |
Green Features | Sustainable packaging |
Product Line | FastRuler |
Product Code | Brand | Size Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size Range | Price | Quantity & Availability | |||||
10410280
![]() |
Thermo Scientific™
SM1233 |
10 to 200 bp |
820.00 NOK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10528740
![]() |
Thermo Scientific™
SM1103 |
50 to 1500 bp |
720.00 NOK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10190740
![]() |
Thermo Scientific™
SM1113 |
100 to 5000 bp |
700.00 NOK
1000µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10669320
![]() |
Thermo Scientific™
SM1123 |
500 to 10000 bp |
722.00 NOK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific FastRuler DNA Ladders is a fast and convenient tool for double-stranded DNA sizing during electrophoresis. It's specifically engineered for rapid sizing and approximate quantification of DNA on both conventional agarose gels and high-throughput E-Gels (in 48- and 96-well formats).
The ladder is a mixture of five blunt-ended chromatography-purified individual DNA fragments. This allows you to achieve precise results faster, saving valuable lab time.
Highlights
- Variety of available ranges (from ultra-low to high- range)
- Fast separation (8 to 14 min)
- Short separation distance (10 to 20 mm)
- Sharp bands, easy-to-remember fragment sizes and quantities
- Ready-to-use: premixed with 6X MassRuler DNA Loading Dye for direct loading
- Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use FastRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
Specifications
Agarose | |
Agarose Gels | |
FastRuler |
Yes | |
Sustainable packaging |
Safety and Handling
missing translation for 'shelfLife' : 24 months when stored at room temperature.