missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM103A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19422-1-AP-20UL
This item is not returnable.
View return policy
Description
FAM103A1, also named as RNMT-activating mini protein, is a 118 amino acid protein, which belongs to the RAM family. FAM103A1 as a component of mRNA cap binding complex localizes in the nucleus. FAM103A1 is required for efficient mRNA cap methylation and regulates RNMT expression by a post-transcriptional stabilizing mechanism.Specifications
FAM103A1 | |
Polyclonal | |
Unconjugated | |
FAM103A1 | |
C15orf18, FAM103A1, HsT19360, Protein FAM103A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
293058, 67148, 83640 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BTL3, Q9CQY2 | |
Fam103a1 | |
FAM103A1 Fusion Protein Ag13749 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction