Learn More
Wheaton™ Extended Controlled Dropper Tip
Shop All Wheaton Science Products Products
Click to view available options
Color:
Natural
Purple
White
Yellow
Description:
Tip size: 13mm
Tip size: 20mm
Tip size: 8mm
Specifications
Specifications
Color | Purple |
Description | Tip size: 20mm |
Material | LDPE |
Drop Size | 41 μL |
Product Type | Extended Controlled Dropper Tip |