missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12723-1-AP-20UL
This item is not returnable.
View return policy
Description
The product of this gene belongs to the SEC15 family. It is highly similar to the protein encoded by Saccharomyces cerevisiae SEC15 gene. This protein is essential for vesicular traffic from the Golgi apparatus to the cell surface in yeast. It is one of the components of a multiprotein complex required for exocytosis. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
EXOC6 | |
Polyclonal | |
Unconjugated | |
EXOC6 | |
EXOC6, EXOC6A, exocyst complex component 6, FLJ1125, SEC15, SEC15 like protein 1, SEC15A, SEC15L, SEC15L1, SEC15L3, Sec15p | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107371, 50556, 54536 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O54923, Q8R313, Q8TAG9 | |
EXOC6 | |
EXOC6 Fusion Protein Ag3418 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction