Learn More
Invitrogen™ EVOS™ Vessel Holder, one Nunc™ or Corning™ T-75 flask
EVOS™ vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems for increased precision in sample alignment
Brand: Invitrogen™ AMEPVH072
This item is not returnable.
View return policy
Description
This particular vessel holder securely holds one NuncÖ or CorningÖ T-75 flask. It was designed for the automated stage of the EVOS™ FL Auto Imaging System but can also be used with the EVOS™ XL or EVOS™ FL imaging systems with stage plate adapter (Cat. No. AMEP4691). It is just one of many easy-to-change vessel holders and stage plates available for this instrument platform. With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.For additional choices, see the EVOS™ Guide to Vessel Holders and Stage Plates.
Explore the entire EVOS™ line of imaging systems and accessories.
Specifications
Vessel Holder | |
one Nunc or Corning T-75 flask | |
1 each |
EVOS | |
Nunc or Corning T-75 Flask | |
Room Temperature |