Learn More
Heidolph™ Evaporating Flask
For use with rotary evaporators
Brand:  Heidolph™ 514-73000-00
947.60 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10457821
  • 974.00 NOK / Each
Only null left
Estimated Shipment: 25-08-2025
Description
Evaporating flasks come with a standard NS 29/32 joint size but are also available in NS 24/29
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
Evaporating Flask
500 mL
115/230 VAC
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Heidolph™ Evaporating Flask > Capacity: 500 mL