Learn More
Memmert™ Entry Port 23 mm for UF260, Custom Position TBC Left Side
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF260F4
2175.97 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 17519145
  • 2235.00 NOK / Each
Only null left
Estimated Shipment: 13-08-2025
Description
  • Entry port with 23 mm diameter
  • Flap closure
  • Custom position TBC in left side
Specifications
Specifications
UF260 Oven
Entry Port
23 mm