missing translation for 'onlineSavingsMsg'
Learn More

(E)-1-Heptene-1,2-diboronic acid bis(pinacol) ester, 98%, Thermo Scientific Chemicals

Product Code. 11305465
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307531-74-4
Molecular Formula C19H36B2O4
Molecular Weight (g/mol) 350.113
MDL Number MFCD02683463
InChI Key KBMWLQPDFRGVTD-PFONDFGASA-N
Synonym 1-cis-1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl heptene, e-1-heptene-1,2-diboronic acid bis pinacol ester, 1,2-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-1-heptene, 4,4,5,5-tetramethyl-2-1e-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl hept-1-en-1-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-e-1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl hept-1-en-2-yl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-tetramethyl-1,3,2-dioxaborolan-2-yl hept-1-en-2-yl-1,3,2-dioxaborolane
PubChem CID 5708398
IUPAC Name 4,4,5,5-tetramethyl-2-[(E)-1-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)hept-1-en-2-yl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C=C(B2OC(C(O2)(C)C)(C)C)CCCCC

Specifications

Boiling Point 108°C (0.08 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 8859280
Formula Weight 350.12
Percent Purity 98%
Chemical Name or Material (E)-1-Heptene-1,2-diboronic acid bis(pinacol) ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.