Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread, and standard ground cone
The threaded tube with thread and standard ground joint allows the bonding of various glass components.
106.76 NOK - 3095.00 NOK
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | ||||||
15395817
|
DWK Life Sciences
248416109 |
GL 18, with Cone NS 14/23 |
1860.00 NOK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
13427467
|
DWK Life Sciences
248406202 |
GL 14, with Cone NS 14/23 | N/A | |||||||
13595370
|
DWK Life Sciences
248407207 |
GL 14, with Cone NS 19/26 |
2320.00 NOK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15222009
|
DWK Life Sciences
248409208 |
GL 14, with Cone NS 29/32 |
3045.00 NOK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15305827
|
DWK Life Sciences
248419106 |
GL 18, with Cone NS 29/32 |
2940.00 NOK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15282009
|
DWK Life Sciences
248429201 |
GL 25, with Cone NS 29/32 |
2790.00 NOK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15292009
|
DWK Life Sciences
248449109 |
GL 32, with Cone NS 29/32 |
3095.00 NOK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15242009
|
DWK Life Sciences
248417105 |
GL 18, with Cone NS 19/26 |
1960.00 NOK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15272009
|
DWK Life Sciences
248428205 |
GL 25, with Cone NS 24/29 |
2555.00 NOK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11639978
|
DWK Life Sciences
248448104 |
GL 32, with Cone NS 24/29 |
2885.00 NOK
Pack of 10 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
12799696
|
DWK Life Sciences
248418101 |
GL 18, with Cone NS 24/29 |
2325.00 NOK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
15656935
|
DWK Life Sciences
248427209 |
GL 25, with Cone NS 19/26 |
2320.00 NOK
Pack of 10 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
||||||
12789696
|
DWK Life Sciences
248408203 |
GL 14, with Cone NS 24/29 |
2710.00 NOK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
Description
This threaded tube has a screw thread and a standard ground joint. Components of an apparatus can be connected via both ends and thus connected to each other.
- Tight closure due to standard ground joint
- With DIN thread
Specifications
10/Pk. |
Product Title
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread, and standard ground cone