Learn More
DWK Life Sciences DURAN™ Short Ground Cone, finished
For connections where space is at a premium.
Brand: DWK Life Sciences 286153606
This item is not returnable.
View return policy
Description
The short-section version of the DURAN™ cores is ideal for use where space is limited and tightness requirements are lower.
- Ground (finer grinding quality, less roughness depth)
- Short grinding for apertures with low space availability and lower tightness requirements
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifications
50/Pk. | |
Installation in glass apparatus and devices | |
13 mm | |
1.5 mm |