Learn More
DWK Life Sciences DURAN™ Receiver Adapter, bent, with standard ground cone
The receiver adapter is suitable for transferring the distillate from the condenser to the receiving flask.
Description
Part of a distillation assembly is, among other things, the receiver adapter, which is connected downstream at the end of the condenser and guides the distillate into the receiving flask.
- Tight closure due to standard ground joint
- Bent
- With standard ground socket
- Fired-on, highly durable white print
Specifications
1 Ea. |