Learn More
DWK Life Sciences DURAN™ Evaporating Dish, with spout
The wide, open shape of the evaporation dish creates a large liquid surface which favours the evaporation of excess solvents.
96.70 - 1060.00
Specificatie
Autoclaveerbaar | Autoclavable |
---|---|
Materiaal | Borosilicate Glass (3.3) |
Voor gebruik met (toepassing) | Evaporation of liquids; Preparation of concentrated solutions; Prevention of solvent condensation on glass walls; Sizes 25, 30, 35 and 45 mL without labelling field |
Aantal per verpakking | 10/Pk. |
Productcode | Merk | Inhoud (metrisch) | Prijs | Hoeveelheid en beschikbaarheid | |||||
---|---|---|---|---|---|---|---|---|---|
Productcode | Merk | Inhoud (metrisch) | Prijs | Hoeveelheid en beschikbaarheid | |||||
11746223
|
DWK Life Sciences
213013202 |
15 mL |
96.70
10 stuks |
|
|||||
11756223
|
DWK Life Sciences
213013408 |
45 mL |
99.45
10 stuks |
|
|||||
11766223
|
DWK Life Sciences
213013802 |
60 mL |
113.00
10 stuks |
|
|||||
11776223
|
DWK Life Sciences
213014104 |
90 mL |
150.00
Pack of 10 |
|
|||||
11786223
![]() |
DWK Life Sciences
213014404 |
170 mL |
131.00
Pack of 10 |
|
|||||
11796223
|
DWK Life Sciences
213014901 |
320 mL |
196.00
Pack of 10 |
![]() |
|||||
11706233
|
DWK Life Sciences
213015409 |
600 mL |
297.00
10 stuks |
|
|||||
11707018
|
DWK Life Sciences
213015906 |
1500 mL |
728.00
10 stuks |
|
|||||
11717018
|
DWK Life Sciences
213016302 |
2500 mL |
1060.00
10 stuks |
|
|||||
Beschrijving
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work.
- Spout for clean pouring
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specificatie
Autoclavable | |
Borosilicate Glass (3.3) | |
Evaporation of liquids; Preparation of concentrated solutions; Prevention of solvent condensation on glass walls; Sizes 25, 30, 35 and 45 mL without labelling field | |
10/Pk. |