Learn More
DWK Life Sciences DURAN™ Drying Tube, bent, with standard ground cone
Filled with a desiccant, the drying tube protects a reaction mixture in a flask from moisture.
329.00 NOK - 393.00 NOK
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
10614182
|
DWK Life Sciences
242620609 |
NS 14/23 |
329.00 NOK
Each |
Estimated Shipment: 13-10-2025
Log in to see stock. |
|||||
10090001
|
DWK Life Sciences
242620909 |
NS 29/32 |
393.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
10400503
|
DWK Life Sciences
242620806 |
NS 24/29 |
387.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
10203221
|
DWK Life Sciences
242620703 |
NS 19/26 |
357.00 NOK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
Description
The drying tube is usually an angled glass tube with a standard ground joint at one end. The other end is open. For filling the drying tube is first stuffed with a little cotton wool, then the drying agent is filled in and finally another piece of cotton wool prevents the drying agent from falling out.
- Tight closure due to standard ground joint
- Bent
- With standard ground socket
- Fired-on, highly durable white print
Specifications
1 Ea. |