Learn More
IKA™ DS 4 Tube Attachment
Brand: IKA™ 0004524300
This item is not returnable.
View return policy
Description
Specifications
30 mL | |
30 mL | |
DS 4 Tube Attachment | |
25 mm |
Product Content Correction
Product Title
IKA™ DS 4 Tube Attachment > Capacity: 30mL